Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF263 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005741.4 | NP_005732.2 | 6 | 1251 | ||
XM_006720831.1 | XP_006720894.1 | 6 | 1319 | |||
XM_006720832.1 | XP_006720895.1 | 5 | 1241 | |||
XM_006720833.1 | XP_006720896.1 | 4 | 1103 | |||
XM_006720834.3 | XP_006720897.1 | 5 | 1374 | |||
XM_011522344.1 | XP_011520646.1 | 6 | 1358 | |||
XM_017022850.1 | XP_016878339.1 | 5 | 797 | |||
GenBank mRNA | AK295350.1 | - | 2 | 117 | ||
AK312421.1 | - | 6 | 1233 | |||
BC008805.2 | - | 6 | 1249 | 3945548 | ||
D88827.1 | - | 6 | 1211 |