Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ALG3 SIRNA Assays ›
Gene Symbol:
ALG3
Gene Name:
ALG3, alpha-1,3- mannosyltransferase
Gene Aliases:
CDG1D, CDGS4, CDGS6, D16Ertd36e, NOT56L, Not56, not
Chromosome Location:
Chr.3: 184242329 - 184249525 on Build GRCh38
UniGene ID:
Hs.478481
Species:
Human
siRNA ID
HSS115536
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ALG3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001006941.2
NP_001006942.1
3
485
NM_005787.5
NP_005778.1
3
412
NR_024533.1
2
312
NR_024534.1
3
375
XM_011512322.1
XP_011510624.1
3
297
XM_011512323.2
XP_011510625.1
3
283
GenBank mRNA
AK289361.1
-
3
441
AK297701.1
-
2
266
AK299575.1
-
2
303
BC002839.2
-
3
387
3636604
BC004313.1
-
3
367
BM920198.1
-
3
341
BX339649.2
-
3
490
KP189279.1
-
3
405
KP189280.1
-
3
368
KP189281.1
-
2
221
KP189282.1
-
3
522
KP189283.1
-
2
305
KP189284.1
-
3
403
KP189285.1
-
3
366
KP189286.1
-
2
221
KP189287.1
-
3
321
KP189288.1
-
2
303
KP189289.1
-
3
402
KP189290.1
-
3
366
KP189291.1
-
2
221
KP189292.1
-
3
321
KP189293.1
-
2
303
KP189294.1
-
3
381
KP189295.1
-
3
381
KP189296.1
-
3
344
KP189297.1
-
3
344
KP189298.1
-
1
95
KP189299.1
-
3
237
KP189300.1
-
2
281
KP189301.1
-
3
344
KP189302.1
-
3
402
KP189303.1
-
3
364
KP189304.1
-
2
221
KP189305.1
-
3
321
KP189306.1
-
2
301
KP189307.1
-
3
401
KP189308.1
-
3
364
KP189309.1
-
2
221
KP189310.1
-
3
321
KP189311.1
-
2
301
KP189312.1
-
3
401
KP189313.1
-
3
401
KP189314.1
-
3
401
KP189315.1
-
3
364
KP189316.1
-
3
364
KP189317.1
-
3
364
KP189318.1
-
2
221
KP189319.1
-
2
221
KP189320.1
-
2
221
KP189321.1
-
3
321
KP189322.1
-
3
321
KP189323.1
-
3
321
KP189324.1
-
2
301
KP189325.1
-
2
301
KP189326.1
-
2
301
KP189327.1
-
2
301
Y09022.1
-
3
412
Back To Top
More Information
Panther Classification:
Molecular Function -
glycosyltransferase
Gene Ontology Categories:
Function(s)
Process(es)
protein glycosylation
dolichol-linked oligosaccharide biosynthetic process
mannosylation
alpha-1,3-mannosyltransferase activity
dol-P-Man:Man(5)GlcNAc(2)-PP-Dol alpha-1,3-mannosyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-d4wl8:80/10.239.162.132:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0