Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPHOSPH9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022782.3 | NP_073619.3 | 10 | 1725 | ||
XM_011537740.2 | XP_011536042.1 | 10 | 1629 | |||
XM_011537741.2 | XP_011536043.1 | 10 | 1629 | |||
XM_017018673.1 | XP_016874162.1 | 10 | 1723 | |||
XM_017018674.1 | XP_016874163.1 | 10 | 1629 | |||
XM_017018675.1 | XP_016874164.1 | 9 | 1531 | |||
XM_017018676.1 | XP_016874165.1 | 10 | 1629 | |||
XM_017018677.1 | XP_016874166.1 | 10 | 1629 | |||
XM_017018678.1 | XP_016874167.1 | 9 | 1449 | |||
XM_017018679.1 | XP_016874168.1 | 11 | 1662 | |||
XM_017018680.1 | XP_016874169.1 | 9 | 1571 | |||
XM_017018681.1 | XP_016874170.1 | 9 | 1484 | |||
GenBank mRNA | AK023016.1 | - | 5 | 448 | ||
AK091181.1 | - | 6 | 1054 | |||
AL096751.1 | - | 4 | 385 | |||
BC130440.1 | - | 6 | 1007 | 40146471 | ||
BC144016.1 | - | 5 | 881 |