Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NME6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308426.1 | NP_001295355.1 | 3 | 490 | ||
NM_001308427.1 | NP_001295356.1 | 3 | 299 | |||
NM_001308428.1 | NP_001295357.1 | 3 | 290 | |||
NM_001308430.1 | NP_001295359.1 | 3 | 273 | |||
NM_001308431.1 | NP_001295360.1 | 3 | 490 | |||
NM_001308433.1 | NP_001295362.1 | 3 | 290 | |||
NM_001308434.1 | NP_001295363.1 | 2 | 393 | |||
NM_001308435.1 | NP_001295364.1 | 3 | 490 | |||
NM_005793.4 | NP_005784.1 | 3 | 273 | |||
XM_017005511.1 | XP_016861000.1 | 3 | 201 | |||
XM_017005513.1 | XP_016861002.1 | 3 | 215 | |||
XM_017005514.1 | XP_016861003.1 | 4 | 472 | |||
XM_017005515.1 | XP_016861004.1 | 3 | 268 | |||
XM_017005516.1 | XP_016861005.1 | 3 | 230 | |||
XM_017005518.1 | XP_016861007.1 | 1 | 41 | |||
GenBank mRNA | AF051941.1 | - | 3 | 206 | ||
AK222555.1 | - | 2 | 172 | |||
AK294809.1 | - | 3 | 246 | |||
AK297364.1 | - | 3 | 199 | |||
AK311152.1 | - | 1 | 41 | |||
BC001808.2 | - | 3 | 220 | 2966629 | ||
BC012828.2 | - | 3 | 220 | 2966629 | ||
BE866839.1 | - | 3 | 241 | |||
BG284082.1 | - | 3 | 219 | |||
BG530435.1 | - | 2 | 95 | |||
CA848473.1 | - | 3 | 269 | |||
CD701581.1 | - | 3 | 243 | |||
CT002144.1 | - | 3 | 490 | |||
DA398008.1 | - | 3 | 246 | |||
HY090220.1 | - | 3 | 243 | |||
U90449.1 | - | 3 | 220 |