Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NME6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308426.1 | NP_001295355.1 | 5, 6 | 732 | ||
NM_001308427.1 | NP_001295356.1 | 5, 6 | 541 | |||
NM_001308428.1 | NP_001295357.1 | 5, 6 | 532 | |||
NM_001308430.1 | NP_001295359.1 | 4, 5 | 475 | |||
NM_001308431.1 | NP_001295360.1 | 4, 5 | 692 | |||
NM_001308433.1 | NP_001295362.1 | 4, 5 | 492 | |||
NM_001308434.1 | NP_001295363.1 | 4, 5 | 635 | |||
NM_005793.4 | NP_005784.1 | 5, 6 | 515 | |||
XM_017005512.1 | XP_016861001.1 | 2, 3 | 1066 | |||
XM_017005513.1 | XP_016861002.1 | 5, 6 | 457 | |||
XM_017005514.1 | XP_016861003.1 | 6, 7 | 714 | |||
XM_017005515.1 | XP_016861004.1 | 4, 5 | 470 | |||
XM_017005517.1 | XP_016861006.1 | 2, 3 | 232 | |||
XM_017005518.1 | XP_016861007.1 | 3, 4 | 361 | |||
GenBank mRNA | AF051941.1 | - | 5, 6 | 448 | ||
AK222555.1 | - | 4, 5 | 414 | |||
AK294809.1 | - | 4, 5 | 448 | |||
AK308970.1 | - | 2, 3 | 219 | |||
BC001808.2 | - | 5, 6 | 462 | 2966629 | ||
BC012828.2 | - | 5, 6 | 462 | 2966629 | ||
BE866839.1 | - | 5, 6 | 484 | |||
BG284082.1 | - | 4, 5 | 421 | |||
CT002144.1 | - | 4, 5 | 692 | |||
DA398008.1 | - | 5, 6 | 488 | |||
U90449.1 | - | 5, 6 | 462 |