Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PATJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176877.2 | NP_795352.2 | 20 | 2683 | ||
XM_005270341.3 | XP_005270398.1 | 20 | 2681 | |||
XM_005270347.1 | XP_005270404.1 | 20 | 2681 | |||
XM_006710278.3 | XP_006710341.1 | 20 | 2681 | |||
XM_011540462.2 | XP_011538764.1 | 20 | 2681 | |||
XM_011540463.2 | XP_011538765.1 | 20 | 2869 | |||
XM_011540464.2 | XP_011538766.1 | 20 | 2681 | |||
XM_011540465.2 | XP_011538767.1 | 20 | 2681 | |||
XM_011540466.2 | XP_011538768.1 | 20 | 2681 | |||
XM_011540467.2 | XP_011538769.1 | 20 | 2681 | |||
XM_011540468.2 | XP_011538770.1 | 20 | 2681 | |||
XM_011540469.2 | XP_011538771.1 | 20 | 2681 | |||
XM_011540470.1 | XP_011538772.1 | 20 | 2681 | |||
XM_016999998.1 | XP_016855487.1 | 20 | 2681 | |||
XM_016999999.1 | XP_016855488.1 | 20 | 2681 | |||
XM_017000000.1 | XP_016855489.1 | 15 | 1927 | |||
GenBank mRNA | AB044807.1 | - | 7 | 962 | ||
AF397170.1 | - | 19 | 2569 | |||
AJ001306.1 | - | 20 | 2683 | |||
AJ224747.1 | - | 20 | 2683 | |||
AJ224748.1 | - | 20 | 2683 | |||
AK304355.1 | - | 15 | 2040 | |||
AK307713.1 | - | 20 | 2680 | |||
BC142661.1 | - | 20 | 2666 | 40148087 | ||
BC142723.1 | - | 20 | 2666 | 40148887 | ||
BC144135.1 | - | 20 | 2635 |