Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPA33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005814.2 | NP_005805.1 | 2 | 509 | ||
XM_017000005.1 | XP_016855494.1 | 3 | 337 | |||
GenBank mRNA | AK312833.1 | - | 2 | 472 | ||
BC069705.1 | - | 2 | 509 | 7262529 | ||
BC069723.1 | - | 2 | 509 | 7262541 | ||
BC069745.1 | - | 2 | 509 | 7262553 | ||
BC069761.1 | - | 2 | 509 | 7262565 | ||
BC069789.1 | - | 2 | 509 | 7262482 | ||
BC074830.2 | - | 2 | 187 | 30915543 | ||
BC074876.2 | - | 2 | 187 | 30915236 | ||
BC107164.1 | - | 2 | 464 | 40034324 | ||
BC107165.1 | - | 2 | 466 | 40034331 | ||
U79725.1 | - | 2 | 509 |