Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPEG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173476.1 | NP_001166947.1 | 4 | 555 | ||
NM_005876.4 | NP_005867.3 | 9 | 2810 | |||
XM_005246237.2 | XP_005246294.1 | 11 | 2538 | |||
XM_005246239.2 | XP_005246296.1 | 11 | 2353 | |||
XM_005246240.2 | XP_005246297.1 | 4 | 555 | |||
XM_005246241.1 | XP_005246298.1 | 4 | 449 | |||
XM_005246242.4 | XP_005246299.1 | 4 | 574 | |||
XM_006712189.3 | XP_006712252.1 | 10 | 2543 | |||
XM_006712193.3 | XP_006712256.1 | 4 | 3674 | |||
XM_011510479.2 | XP_011508781.1 | 10 | 2976 | |||
XM_011510483.2 | XP_011508785.2 | 10 | 2573 | |||
XM_017003157.1 | XP_016858646.1 | 10 | 2573 | |||
XM_017003158.1 | XP_016858647.1 | 6 | 2149 | |||
XM_017003159.1 | XP_016858648.1 | 7 | 2332 | |||
XM_017003161.1 | XP_016858650.1 | 7 | 2332 | |||
XM_017003162.1 | XP_016858651.1 | 7 | 2332 | |||
GenBank mRNA | AK055387.1 | - | 5 | 1163 | ||
AK126500.1 | - | 10 | 2998 | |||
AK128689.1 | - | 9 | 2808 | |||
AK129604.1 | - | 3 | 1012 | |||
AK289531.1 | - | 11 | 2335 | |||
AY603755.1 | - | 8 | 2423 | |||
BC006346.2 | - | 4 | 397 | |||
CR542201.1 | - | 3 | 263 | |||
DA063220.1 | - | 4 | 556 | |||
U57099.1 | - | 4 | 388 |