Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRVI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098579.2 | NP_001092049.2 | 20 | 2673 | ||
NM_001100163.2 | NP_001093633.1 | 21 | 2951 | |||
NM_001100167.2 | NP_001093637.1 | 21 | 2668 | |||
NM_001206880.1 | NP_001193809.1 | 20 | 2386 | |||
NM_001206881.1 | NP_001193810.1 | 19 | 2793 | |||
NM_130385.3 | NP_569056.4 | 21 | 3004 | |||
GenBank mRNA | AF081249.1 | - | 19 | 2689 | ||
AF081250.1 | - | 19 | 2417 | |||
AK127209.1 | - | 21 | 2646 | |||
AK296336.1 | - | 21 | 2768 | |||
AK300358.1 | - | 20 | 2658 | |||
AK304702.1 | - | 20 | 2020 | |||
BC117127.1 | - | 21 | 2541 | 40125678 |