Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006993.2 | NP_008924.1 | 4, 5 | 441 | ||
GenBank mRNA | AF081280.1 | - | 4, 5 | 429 | ||
AY049737.1 | - | 4, 5 | 433 | |||
BC041067.1 | - | 4, 5 | 422 | 5473410 | ||
BC054868.1 | - | 4, 5 | 427 | 6496849 | ||
BM739536.1 | - | 4, 5 | 441 | |||
BM765409.1 | - | 4, 5 | 441 |