Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACNG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006078.3 | NP_006069.1 | 1, 2 | 271 | ||
GenBank mRNA | AF096322.1 | - | 1, 2 | 202 | ||
BC069612.1 | - | 1, 2 | 271 | 7262295 | ||
BC112297.1 | - | 1, 2 | 262 | 8327765 | ||
BC112299.1 | - | 1, 2 | 262 | 8327767 | ||
BE670632.1 | - | 1, 2 | 485 | |||
CR456414.1 | - | 1, 2 | 237 |