Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACNG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006078.3 | NP_006069.1 | 4 | 534 | ||
XM_017028531.1 | XP_016884020.1 | 3 | 331 | |||
GenBank mRNA | AF096322.1 | - | 4 | 465 | ||
BC069612.1 | - | 4 | 534 | 7262295 | ||
BC112297.1 | - | 4 | 525 | 8327765 | ||
BC112299.1 | - | 4 | 525 | 8327767 | ||
CR456414.1 | - | 4 | 500 |