Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBB4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289123.1 | NP_001276052.1 | 5 | 911 | ||
NM_001289127.1 | NP_001276056.1 | 5 | 893 | |||
NM_001289129.1 | NP_001276058.1 | 5 | 841 | |||
NM_001289130.1 | NP_001276059.1 | 4 | 950 | |||
NM_001289131.1 | NP_001276060.1 | 4 | 917 | |||
NM_006087.3 | NP_006078.2 | 4 | 993 | |||
GenBank mRNA | AK095202.1 | - | 3 | 1250 | ||
AK293503.1 | - | 4 | 642 | |||
BC006570.2 | - | 4 | 702 | 3603726 | ||
BC013683.1 | - | 4 | 728 | 4155617 |