Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD2BP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243646.1 | NP_001230575.1 | 3 | 580 | ||
NM_006110.2 | NP_006101.1 | 4 | 503 | |||
GenBank mRNA | AF104222.1 | - | 4 | 447 | ||
AK315708.1 | - | 4 | 388 | |||
BC000495.2 | - | 4 | 402 | 2820690 | ||
BC001947.1 | - | 4 | 402 | 2820690 | ||
BC013854.1 | - | 3 | 580 | |||
KU178518.1 | - | 3 | 327 |