Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PPIE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195007.1 | NP_001181936.1 | 5 | 316 | ||
NM_001319293.1 | NP_001306222.1 | 5 | 316 | |||
NM_006112.3 | NP_006103.1 | 5 | 316 | |||
NM_203456.2 | NP_982281.1 | 5 | 316 | |||
NR_036543.1 | 5 | 308 | ||||
NR_036544.1 | 5 | 308 | ||||
XM_006710289.2 | XP_006710352.1 | 5 | 271 | |||
XM_006710290.3 | XP_006710353.1 | 5 | 271 | |||
XM_011540501.1 | XP_011538803.1 | 5 | 418 | |||
XM_017000051.1 | XP_016855540.1 | 5 | 267 | |||
XM_017000052.1 | XP_016855541.1 | 5 | 418 | |||
GenBank mRNA | AF042385.1 | - | 5 | 320 | ||
AF042386.1 | - | 5 | 320 | |||
AF104012.1 | - | 5 | 271 | |||
AF104013.1 | - | 5 | 271 | |||
AK094660.1 | - | 4 | 1543 | |||
AK130628.1 | - | 5 | 269 | |||
AK293094.1 | - | 5 | 267 | |||
AK304694.1 | - | 5 | 263 | |||
AK313666.1 | - | 5 | 284 | |||
AL833576.1 | - | 5 | 376 | |||
BC004898.2 | - | 5 | 266 | 3545997 | ||
BC008451.1 | - | 5 | 279 | 4134572 | ||
BC107736.1 | - | 5 | 275 | 4809032 | ||
BU160972.1 | - | 5 | 245 | |||
BU179717.1 | - | 303 | ||||
BU196539.1 | - | 5 | 303 | |||
CT002466.1 | - | 5 | 257 | |||
DQ160195.1 | - | 5 | 260 |