Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC30A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006345.3 | NP_006336.3 | 17 | 1783 | ||
XM_017007654.1 | XP_016863143.1 | 17 | 1817 | |||
GenBank mRNA | AF006621.2 | - | 17 | 1757 | ||
AK225470.1 | - | 17 | 1720 | |||
AK299917.1 | - | 18 | 1877 | |||
AK312838.1 | - | 17 | 1783 | |||
AY319413.1 | - | 18 | 1662 | |||
AY594282.1 | - | 17 | 1637 | |||
BC000240.1 | - | 17 | 1745 | |||
BC007732.2 | - | 17 | 1744 | 4109702 | ||
BC016949.1 | - | 17 | 1762 | 3850521 | ||
BC022981.1 | - | 13 | 1174 | |||
KU178526.1 | - | 17 | 1637 | |||
KU178527.1 | - | 17 | 1702 |