Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SEMA4D SIRNA Assays ›
Gene Symbol:
SEMA4D
Gene Name:
semaphorin 4D
Gene Aliases:
C9orf164, CD100, M-sema-G, SEMAJ, coll-4
Chromosome Location:
Chr.9: 89360791 - 89498065 on Build GRCh38
UniGene ID:
Hs.494406
Species:
Human
siRNA ID
HSS116125
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SEMA4D
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001142287.1
NP_001135759.1
13
1635
NM_006378.3
NP_006369.3
13
1635
XM_005251654.3
XP_005251711.1
11
1469
XM_011518123.2
XP_011516425.1
15
1719
XM_011518124.2
XP_011516426.1
15
1678
XM_011518125.1
XP_011516427.1
13
1606
XM_011518127.2
XP_011516429.1
14
1634
XM_011518128.2
XP_011516430.1
13
1559
XM_011518129.1
XP_011516431.1
12
1488
XM_011518130.2
XP_011516432.1
12
1723
XM_011518131.2
XP_011516433.1
11
1661
XM_011518133.2
XP_011516435.1
12
1585
XM_011518134.2
XP_011516436.1
10
1526
XM_017014193.1
XP_016869682.1
14
3329
XM_017014194.1
XP_016869683.1
14
1603
XM_017014195.1
XP_016869684.1
13
1540
XM_017014196.1
XP_016869685.1
13
1877
XM_017014197.1
XP_016869686.1
12
1761
XM_017014198.1
XP_016869687.1
11
1698
XM_017014199.1
XP_016869688.1
15
1678
GenBank mRNA
AB210030.1
-
11
1840
AK293749.1
-
13
1745
AK316107.1
-
13
1829
BC054500.1
-
13
1635
5551182
BX648216.1
-
11
1427
U60800.1
-
9
1150
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane-bound signaling molecule
intercellular signal molecule
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
neural crest cell migration
positive regulation of protein phosphorylation
immune response
cell adhesion
negative regulation of cell adhesion
regulation of cell shape
negative regulation of alkaline phosphatase activity
positive regulation of phosphatidylinositol 3-kinase signaling
positive regulation of cell migration
regulation of cell projection organization
negative regulation of apoptotic process
positive regulation of GTPase activity
ossification involved in bone maturation
negative regulation of osteoblast differentiation
positive regulation of collateral sprouting
regulation of dendrite morphogenesis
negative regulation of axon extension involved in axon guidance
positive regulation of peptidyl-tyrosine phosphorylation
negative regulation of peptidyl-tyrosine phosphorylation
negative chemotaxis
leukocyte aggregation
semaphorin-plexin signaling pathway
semaphorin-plexin signaling pathway involved in bone trabecula morphogenesis
receptor activity
transmembrane signaling receptor activity
receptor binding
protein binding
semaphorin receptor binding
neuropilin binding
chemorepellent activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-l9dfq:80/10.239.161.188:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0