Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARFGEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006421.4 | NP_006412.2 | 21 | 3433 | ||
XM_005251134.4 | XP_005251191.1 | 22 | 3412 | |||
XM_005251135.3 | XP_005251192.1 | 21 | 3683 | |||
XM_005251136.3 | XP_005251193.1 | 21 | 3683 | |||
XM_011517442.2 | XP_011515744.1 | 22 | 3719 | |||
GenBank mRNA | AB209324.1 | - | 11 | 1478 | ||
AF084520.1 | - | 21 | 3043 | |||
AF111162.1 | - | 21 | 3184 | |||
AK025637.1 | - | 2 | 146 |