Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TACC2 SIRNA Assays ›
Gene Symbol:
TACC2
Gene Name:
transforming acidic coiled-coil containing protein 2
Gene Aliases:
AZU-1, ECTACC
Chromosome Location:
Chr.10: 121989174 - 122254545 on Build GRCh38
UniGene ID:
Hs.501252
Species:
Human
siRNA ID
HSS116289
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TACC2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001291876.1
NP_001278805.1
18
8568
NM_001291877.1
NP_001278806.1
18
8715
NM_001291878.1
NP_001278807.1
15
3253
NM_001291879.1
NP_001278808.1
15
2926
NM_006997.3
NP_008928.1
15
3307
NM_206860.2
NP_996742.1
14
3217
NM_206861.2
NP_996743.1
18
3372
NM_206862.3
NP_996744.3
21
8934
XM_005269388.1
XP_005269445.1
22
8851
XM_005269389.4
XP_005269446.1
22
9375
XM_005269390.1
XP_005269447.1
23
8794
XM_005269391.1
XP_005269448.1
21
8791
XM_005269392.1
XP_005269449.1
21
8761
XM_005269393.1
XP_005269450.1
22
8758
XM_005269394.1
XP_005269451.1
21
8725
XM_005269395.1
XP_005269452.1
21
8716
XM_005269396.1
XP_005269453.1
20
8620
XM_005269397.1
XP_005269454.1
20
8590
XM_005269399.1
XP_005269456.1
19
8392
XM_006717548.1
XP_006717611.1
22
8746
XM_006717552.1
XP_006717615.1
21
8563
XM_006717553.1
XP_006717616.1
20
8527
XM_006717554.1
XP_006717617.1
20
8515
XM_011539126.1
XP_011537428.1
22
8848
XM_011539128.1
XP_011537430.1
20
3424
XM_011539129.1
XP_011537431.1
19
3289
XM_011539130.1
XP_011537432.1
19
3196
XM_011539131.1
XP_011537433.1
15
3262
XM_011539132.1
XP_011537434.1
16
3205
XM_011539133.1
XP_011537435.1
14
2879
XM_011539134.1
XP_011537436.1
14
3150
XM_017015484.1
XP_016870973.1
22
8839
XM_017015485.1
XP_016870974.1
22
8704
XM_017015486.1
XP_016870975.1
21
8668
XM_017015487.1
XP_016870976.1
21
8656
XM_017015488.1
XP_016870977.1
20
8485
XM_017015489.1
XP_016870978.1
19
8407
XM_017015491.1
XP_016870980.1
15
3402
XM_017015492.1
XP_016870981.1
15
3309
XM_017015493.1
XP_016870982.1
14
3067
XM_017015494.1
XP_016870983.1
13
3078
XM_017015495.1
XP_016870984.1
13
2938
XM_017015496.1
XP_016870985.1
16
3203
XM_017015497.1
XP_016870986.1
16
3462
XM_017015498.1
XP_016870987.1
15
3327
XM_017015499.1
XP_016870988.1
17
3296
GenBank mRNA
AF095791.2
-
15
2894
AF176646.1
-
14
3052
AF220152.2
-
11
2740
AF528098.1
-
18
3353
AF528099.1
-
21
8915
AK027066.1
-
11
1846
AK056743.1
-
2
1163
AK075097.1
-
11
1204
AK095371.1
-
2
2352
AL713712.1
-
11
2152
AL833304.1
-
20
8033
BC000999.2
-
4
301
BC039311.1
-
14
3217
BC141837.1
-
16
2905
BC142673.1
-
16
2905
BC144599.1
-
18
8683
9053129
BC144600.1
-
18
8536
9053130
BC150627.1
-
18
8536
9056997
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
cell proliferation
cerebral cortex development
nuclear hormone receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-r4vqj:80/10.239.163.135:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0