Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SORBS1 SIRNA Assays ›
Gene Symbol:
SORBS1
Gene Name:
sorbin and SH3 domain containing 1
Gene Aliases:
CAP, FLAF2, R85FL, SH3D5, SH3P12, SORB1
Chromosome Location:
Chr.10: 95311773 - 95561420 on Build GRCh38
UniGene ID:
Hs.38621
Species:
Human
siRNA ID
HSS116290
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SORBS1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001034954.1
NP_001030126.1
23, 24
2435
NM_001034955.1
NP_001030127.1
23, 24
2501
NM_001034956.1
NP_001030128.1
20, 21
1988
NM_001034957.1
NP_001030129.1
15, 16
1553
NM_001290294.1
NP_001277223.1
23, 24
2484
NM_001290295.1
NP_001277224.1
18, 19
1767
NM_001290296.1
NP_001277225.1
21, 22
2345
NM_001290297.1
NP_001277226.1
18, 19
1766
NM_001290298.1
NP_001277227.1
19, 20
1871
NM_006434.2
NP_006425.2
14, 15
1385
NM_015385.3
NP_056200.1
18, 19
1968
NM_024991.2
NP_079267.1
19, 20
1863
XM_006717589.1
XP_006717652.1
23, 24
2485
XM_006717593.1
XP_006717656.1
17, 18
1718
XM_011539140.1
XP_011537442.1
25, 26
3229
XM_011539150.1
XP_011537452.1
24, 25
3154
XM_011539155.1
XP_011537457.1
19, 20
2804
XM_011539167.1
XP_011537469.1
18, 19
2087
XM_017015500.1
XP_016870989.1
20, 21
2734
XM_017015501.1
XP_016870990.1
24, 25
3154
XM_017015502.1
XP_016870991.1
23, 24
3061
XM_017015503.1
XP_016870992.1
22, 23
2986
XM_017015504.1
XP_016870993.1
23, 24
2143
XM_017015505.1
XP_016870994.1
21, 22
2902
XM_017015506.1
XP_016870995.1
22, 23
2068
XM_017015507.1
XP_016870996.1
22, 23
2893
XM_017015508.1
XP_016870997.1
21, 22
1975
XM_017015509.1
XP_016870998.1
21, 22
1997
XM_017015510.1
XP_016870999.1
20, 21
2734
XM_017015511.1
XP_016871000.1
20, 21
1891
XM_017015512.1
XP_016871001.1
18, 19
1802
XM_017015513.1
XP_016871002.1
18, 19
1793
XM_017015514.1
XP_016871003.1
18, 19
1709
XM_017015515.1
XP_016871004.1
22, 23
2344
XM_017015516.1
XP_016871005.1
21, 22
2317
XM_017015517.1
XP_016871006.1
21, 22
2260
XM_017015518.1
XP_016871007.1
21, 22
2273
XM_017015519.1
XP_016871008.1
20, 21
2246
XM_017015520.1
XP_016871009.1
20, 21
2185
XM_017015521.1
XP_016871010.1
20, 21
2198
XM_017015522.1
XP_016871011.1
19, 20
2158
XM_017015523.1
XP_016871012.1
19, 20
2114
XM_017015524.1
XP_016871013.1
18, 19
2087
XM_017015525.1
XP_016871014.1
22, 23
2072
XM_017015526.1
XP_016871015.1
21, 22
1975
XM_017015527.1
XP_016871016.1
21, 22
1997
XM_017015528.1
XP_016871017.1
20, 21
1948
XM_017015529.1
XP_016871018.1
20, 21
1900
XM_017015530.1
XP_016871019.1
20, 21
1891
XM_017015531.1
XP_016871020.1
20, 21
1904
XM_017015532.1
XP_016871021.1
19, 20
1873
XM_017015533.1
XP_016871022.1
19, 20
1877
XM_017015534.1
XP_016871023.1
20, 21
1807
XM_017015535.1
XP_016871024.1
19, 20
1829
XM_017015536.1
XP_016871025.1
19, 20
1820
XM_017015537.1
XP_016871026.1
18, 19
1789
XM_017015538.1
XP_016871027.1
19, 20
1780
XM_017015539.1
XP_016871028.1
18, 19
1802
XM_017015540.1
XP_016871029.1
18, 19
1793
GenBank mRNA
AB037717.1
-
19, 20
1795
AF136380.1
-
13, 14
1340
AF136381.1
-
20, 21
2300
AF330623.1
-
17, 18
1721
AF330624.1
-
3, 4
419
AF356525.1
-
23, 24
2446
AF356526.1
-
23, 24
2512
AF356527.1
-
20, 21
1999
AJ489942.1
-
18, 19
1826
AK294924.1
-
7, 8
745
AK300407.1
-
18, 19
1720
AK304130.1
-
7, 8
808
AL117472.1
-
18, 19
1926
AM260536.1
-
23, 24
2442
BC152463.1
-
19, 20
1795
8862599
BX640814.1
-
7, 8
1798
U70668.1
-
1, 2
137
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
muscle contraction
cell-matrix adhesion
insulin receptor signaling pathway
positive regulation of signal transduction
glucose transport
cellular response to insulin stimulus
stress fiber assembly
positive regulation of glycogen biosynthetic process
positive regulation of glucose import
positive regulation of lipid biosynthetic process
focal adhesion assembly
positive regulation of establishment of protein localization to plasma membrane
actin binding
SH3/SH2 adaptor activity
insulin receptor binding
protein binding
cytoskeletal protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0