Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COLEC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006438.4 | NP_006429.2 | 1 | 186 | ||
GenBank mRNA | AB002631.1 | - | 1 | 94 | ||
AK291791.1 | - | 1 | 186 | |||
AY359038.1 | - | 1 | 110 | |||
BC103815.1 | - | 1 | 130 | 40000432 | ||
BC103816.1 | - | 1 | 130 | 40000433 |