Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CAMKK2 SIRNA Assays ›
Gene Symbol:
CAMKK2
Gene Name:
calcium/calmodulin dependent protein kinase kinase 2
Gene Aliases:
CAMKK, CAMKKB
Chromosome Location:
Chr.12: 121237692 - 121298308 on Build GRCh38
UniGene ID:
Hs.297343
Species:
Human
siRNA ID
HSS116423
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CAMKK2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001270485.1
NP_001257414.1
7, 8
965
NM_001270486.1
NP_001257415.1
6, 7
849
NM_006549.3
NP_006540.3
7, 8
1619
NM_153499.2
NP_705719.2
7, 8
1619
NM_153500.1
NP_705720.1
7, 8
1619
NM_172214.2
NP_757363.1
7, 8
1619
NM_172215.2
NP_757364.1
7, 8
1619
NM_172216.1
NP_757365.1
7, 8
1619
NM_172226.2
NP_757380.1
7, 8
965
XM_005253822.3
XP_005253879.1
7, 8
921
XM_005253823.1
XP_005253880.1
7, 8
921
XM_005253824.3
XP_005253881.1
7, 8
921
XM_011537763.1
XP_011536065.1
7, 8
921
XM_011537764.2
XP_011536066.1
7, 8
967
XM_017018694.1
XP_016874183.1
8, 9
975
XM_017018695.1
XP_016874184.1
8, 9
1184
XM_017018696.1
XP_016874185.1
8, 9
1026
XM_017018697.1
XP_016874186.1
8, 9
1048
XM_017018698.1
XP_016874187.1
7, 8
972
XM_017018699.1
XP_016874188.1
8, 9
975
XM_017018700.1
XP_016874189.1
8, 9
975
XM_017018701.1
XP_016874190.1
7, 8
972
XM_017018702.1
XP_016874191.1
7, 8
1130
XM_017018703.1
XP_016874192.1
8, 9
975
XM_017018704.1
XP_016874193.1
7, 8
972
XM_017018705.1
XP_016874194.1
8, 9
975
XM_017018706.1
XP_016874195.1
7, 8
967
XM_017018707.1
XP_016874196.1
6, 7
890
XM_017018708.1
XP_016874197.1
7, 8
967
GenBank mRNA
AB018330.2
-
6, 7
795
AB081336.1
-
6, 7
822
AB081337.1
-
6, 7
822
AF101264.1
-
6, 7
442
AF140507.1
-
7, 8
903
AF287630.1
-
7, 8
1613
AF287631.1
-
7, 8
1613
AF321385.1
-
7, 8
1369
AF321386.1
-
7, 8
1369
AF321387.1
-
7, 8
1369
AF321388.1
-
7, 8
1369
AK024748.1
-
6, 7
605
AK292072.1
-
7, 8
1113
AK300865.1
-
8, 9
865
AL834322.1
-
1, 2
218
BC000318.2
-
2, 3
94
BC026060.2
-
7, 8
898
3842706
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
protein phosphorylation
peptidyl-tyrosine phosphorylation
calcium-mediated signaling
regulation of protein kinase activity
positive regulation of transcription, DNA-templated
protein autophosphorylation
calmodulin-dependent protein kinase activity
protein tyrosine kinase activity
calcium ion binding
calmodulin binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-6lbnp:80/10.239.161.162:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline