Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256725.1 | NP_001243654.1 | 6 | 920 | ||
NM_001256726.1 | NP_001243655.1 | 6 | 920 | |||
NM_001256727.1 | NP_001243656.1 | 6 | 805 | |||
NM_001256728.1 | NP_001243657.1 | 5 | 730 | |||
NM_006590.3 | NP_006581.2 | 6 | 920 | |||
NR_046347.1 | 6 | 808 | ||||
XM_006711922.1 | XP_006711985.1 | 6 | 788 | |||
XM_006711923.2 | XP_006711986.1 | 6 | 867 | |||
XM_011532487.1 | XP_011530789.1 | 6 | 873 | |||
XM_011532488.1 | XP_011530790.1 | 6 | 806 | |||
XM_017003182.1 | XP_016858671.1 | 6 | 877 | |||
GenBank mRNA | AF130096.1 | - | 5 | 509 | ||
AF132955.1 | - | 6 | 825 | |||
AF353989.1 | - | 6 | 827 | |||
AK001772.1 | - | 2 | 213 | |||
AK056334.1 | - | 6 | 789 | |||
AK129872.1 | - | 7 | 801 | |||
AK222852.1 | - | 6 | 823 | |||
AK289451.1 | - | 6 | 874 | |||
AK295257.1 | - | 5 | 728 | |||
AK302236.1 | - | 6 | 782 | |||
AL117416.1 | - | 4 | 487 | |||
AL519827.3 | - | 6 | 870 | |||
BC001384.2 | - | 6 | 809 | |||
BC067273.1 | - | 6 | 848 | 6503509 | ||
BX462623.2 | - | 6 | 854 |