Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTMR11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145862.1 | NP_001139334.1 | 6 | 759 | ||
NM_181873.3 | NP_870988.2 | 5 | 501 | |||
XM_006711137.1 | XP_006711200.1 | 5 | 478 | |||
XM_011509099.2 | XP_011507401.1 | 6 | 652 | |||
GenBank mRNA | AK097000.1 | - | 5 | 478 | ||
AK296094.1 | - | 5 | 598 | |||
AK298775.1 | - | 6 | 728 | |||
AK304691.1 | - | 5 | 575 | |||
AK304820.1 | - | 6 | 729 | |||
BC032655.1 | - | 5 | 440 | |||
BC064372.1 | - | 6 | 731 | |||
BC094756.1 | - | 6 | 740 | |||
BC100818.2 | - | 5 | 419 | |||
BC100819.2 | - | 5 | 419 | |||
BC100820.1 | - | 5 | 419 | |||
BC100821.2 | - | 5 | 419 | |||
U78556.1 | - | 6 | 694 | |||
U78557.1 | - | 6 | 694 |