- Order Status
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: TXNL4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303471.2 | NP_001290400.1 | 1 | 323 | ||
NM_006701.4 | NP_006692.1 | 1 | 323 | |||
NR_131175.1 | 1 | 323 | ||||
NR_131176.1 | 1 | 323 | ||||
NR_131177.1 | 1 | 323 | ||||
GenBank mRNA | AF023611.1 | - | 1 | 249 | ||
AF023612.1 | - | 1 | 249 | |||
AF146373.1 | - | 1 | 123 | |||
AK309098.1 | - | 1 | 240 | |||
AK314901.1 | - | 1 | 275 | |||
AL522879.3 | - | 1 | 312 | |||
AW796546.1 | - | 1 | 57 | |||
BC001046.2 | - | 1 | 263 | 2823293 | ||
BC019272.1 | - | 1 | 263 | 2823293 | ||
BE385528.1 | - | 1 | 259 | |||
BF972457.1 | - | 1 | 324 | |||
BU553165.1 | - | 267 |