Gene Symbol: TXNL4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303471.2 | NP_001290400.1 | 4 | 694 | ||
NM_001305557.1 | NP_001292486.1 | 3 | 521 | |||
NM_001305563.1 | NP_001292492.1 | 4 | 334 | |||
NM_001305564.1 | NP_001292493.1 | 3 | 277 | |||
NM_006701.4 | NP_006692.1 | 3 | 545 | |||
NR_131175.1 | 5 | 764 | ||||
NR_131176.1 | 4 | 615 | ||||
NR_131177.1 | 2 | 441 | ||||
GenBank mRNA | AA923682.1 | - | 1 | 251 | ||
AF023611.1 | - | 3 | 471 | |||
AF023612.1 | - | 4 | 541 | |||
AF146373.1 | - | 3 | 345 | |||
AK308703.1 | - | 3 | 257 | |||
AK309098.1 | - | 4 | 611 | |||
AK314901.1 | - | 3 | 497 | |||
AL522879.3 | - | 5 | 753 | |||
AV729855.2 | - | 3 | 277 | |||
AW796546.1 | - | 2 | 175 | |||
BC001046.2 | - | 3 | 485 | 2823293 | ||
BC019272.1 | - | 3 | 485 | 2823293 | ||
BE385528.1 | - | 3 | 481 | |||
BF972457.1 | - | 3 | 546 | |||
BM922767.1 | - | 3 | 522 | |||
BQ011103.1 | - | 1 | 270 |