Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUGT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130912.2 | NP_001124384.1 | 13 | 1179 | ||
NM_001320831.1 | NP_001307760.1 | 13 | 1211 | |||
NM_006704.4 | NP_006695.1 | 12 | 1083 | |||
XM_011534891.2 | XP_011533193.1 | 13 | 1274 | |||
XM_011534893.2 | XP_011533195.1 | 12 | 992 | |||
XM_017020365.1 | XP_016875854.1 | 13 | 1164 | |||
XM_017020366.1 | XP_016875855.1 | 13 | 1039 | |||
XM_017020367.1 | XP_016875856.1 | 13 | 1073 | |||
XM_017020368.1 | XP_016875857.1 | 13 | 1085 | |||
GenBank mRNA | AF132856.1 | - | 12 | 853 | ||
AJ344097.1 | - | 12 | 946 | |||
AK291402.1 | - | 13 | 996 | |||
AK292128.1 | - | 12 | 943 | |||
AK302363.1 | - | 13 | 1058 | |||
AY271314.1 | - | 13 | 1016 | |||
AY321358.1 | - | 13 | 1031 | |||
BC000911.1 | - | 12 | 908 | 2985858 | ||
BT009798.1 | - | 12 | 853 |