Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMPDL3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286138.1 | NP_001273067.1 | 3 | 568 | ||
NM_006714.4 | NP_006705.1 | 4 | 782 | |||
XM_005266797.4 | XP_005266854.1 | 4 | 489 | |||
XM_005266799.2 | XP_005266856.1 | 4 | 782 | |||
GenBank mRNA | AK000184.1 | - | 4 | 555 | ||
AK301365.1 | - | 3 | 291 | |||
BC018999.2 | - | 4 | 552 | 3138813 |