Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SF3A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320830.1 | NP_001307759.1 | 6 | 567 | ||
NM_006802.3 | NP_006793.1 | 8 | 726 | |||
GenBank mRNA | AK128438.1 | - | 8 | 1535 | ||
AK222554.1 | - | 8 | 602 | |||
AK291181.1 | - | 8 | 607 | |||
AK292116.1 | - | 8 | 637 | |||
AK301424.1 | - | 6 | 444 | |||
BC002395.2 | - | 8 | 619 | 2821350 | ||
BC011523.1 | - | 8 | 683 | 3867442 | ||
U08815.1 | - | 8 | 589 |