Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006833.4 | NP_006824.2 | 7 | 617 | ||
GenBank mRNA | AK312024.1 | - | 7 | 608 | ||
BC002520.2 | - | 7 | 591 | 3140042 | ||
BI113801.1 | - | 7 | 619 | |||
CB990467.1 | - | 7 | 641 | |||
U70735.1 | - | 7 | 539 |