Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100169.1 | NP_001093639.1 | 14, 15 | 2057 | ||
NM_001100170.1 | NP_001093640.1 | 14, 15 | 2027 | |||
NM_006839.2 | NP_006830.2 | 14, 15 | 2060 | |||
XM_005264110.3 | XP_005264167.1 | 14, 15 | 1696 | |||
XM_005264113.1 | XP_005264170.1 | 13, 14 | 1632 | |||
XM_005264114.1 | XP_005264171.1 | 13, 14 | 1603 | |||
XM_011532497.2 | XP_011530799.1 | 14, 15 | 1720 | |||
XM_017003194.1 | XP_016858683.1 | 14, 15 | 1728 | |||
XM_017003195.1 | XP_016858684.1 | 14, 15 | 1752 | |||
GenBank mRNA | AK225780.1 | - | 14, 15 | 1694 | ||
AK296676.1 | - | 14, 15 | 1666 | |||
AK299014.1 | - | 13, 14 | 1582 | |||
AK299596.1 | - | 12, 13 | 1437 | |||
AK300017.1 | - | 12, 13 | 1403 | |||
AK312251.1 | - | 14, 15 | 1710 | |||
AY232292.1 | - | 14, 15 | 1690 | |||
AY921637.1 | - | 14, 15 | 1708 | |||
BC002412.2 | - | 14, 15 | 1701 | 2964676 | ||
BC027458.1 | - | 3, 4 | 413 | |||
BX537369.1 | - | 14, 15 | 1731 | |||
CR456793.1 | - | 14, 15 | 1672 | |||
CR749590.1 | - | 9, 10 | 1104 | |||
D21092.1 | - | 12, 13 | 1562 | |||
D21093.1 | - | 3, 4 | 324 | |||
D21094.1 | - | 14, 15 | 1757 | |||
L42572.1 | - | 14, 15 | 1764 |