Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297655.1 | NP_001284584.1 | 14 | 1377 | ||
NM_001297656.1 | NP_001284585.1 | 14 | 1618 | |||
NM_001297657.1 | NP_001284586.1 | 14 | 1305 | |||
NM_006845.3 | NP_006836.2 | 15 | 1500 | |||
XM_011540540.2 | XP_011538842.1 | 15 | 1747 | |||
XM_011540541.2 | XP_011538843.1 | 14 | 1688 | |||
GenBank mRNA | AB264115.1 | - | 15 | 1500 | ||
AK290688.1 | - | 15 | 1426 | |||
AK300756.1 | - | 14 | 1334 | |||
AK302267.1 | - | 14 | 1273 | |||
AK312606.1 | - | 15 | 1456 | |||
AY026505.1 | - | 14 | 1571 | |||
BC008764.2 | - | 15 | 1439 | 3347571 | ||
BC014924.1 | - | 15 | 1437 | 3909438 | ||
BC032038.1 | - | 7 | 615 | |||
BT006759.1 | - | 15 | 1385 | |||
CR450302.1 | - | 15 | 1385 | |||
U63743.1 | - | 15 | 1439 |