Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLIPR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006851.2 | NP_006842.2 | 2 | 524 | ||
XM_011537775.2 | XP_011536077.1 | 2 | 503 | |||
XM_017018721.1 | XP_016874210.1 | 2 | 404 | |||
GenBank mRNA | AF400440.1 | - | 2 | 404 | ||
AK304847.1 | - | 2 | 504 | |||
AK313449.1 | - | 2 | 390 | |||
BC012510.1 | - | 2 | 440 | 4453841 | ||
CB123261.1 | - | 2 | 577 | |||
EF667354.1 | - | 2 | 376 | |||
U16307.3 | - | 2 | 460 | |||
X91911.1 | - | 2 | 504 |