Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OGFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007346.2 | NP_031372.2 | 6 | 620 | ||
GenBank mRNA | AF109134.1 | - | 6 | 614 | ||
AF172449.1 | - | 6 | 618 | |||
AF172450.1 | - | 6 | 618 | |||
AF172451.1 | - | 6 | 618 | |||
AF172452.1 | - | 6 | 618 | |||
AF172453.1 | - | 618 | ||||
AK022234.1 | - | 6 | 610 | |||
AK024485.1 | - | 4 | 2706 | |||
AK314696.1 | - | 6 | 612 | |||
BC008768.2 | - | 6 | 596 | 3050950 | ||
BC014137.2 | - | 6 | 595 | 4643904 | ||
BC032666.1 | - | 6 | 575 | 5552531 | ||
BM551600.1 | - | 6 | 620 |