Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM115 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007024.4 | NP_008955.1 | 1 | 805 | ||
GenBank mRNA | AK314166.1 | - | 1 | 812 | ||
BC011948.1 | - | 1 | 769 | 4178931 | ||
BC017367.2 | - | 1 | 759 | 3010216 | ||
CR457020.1 | - | 1 | 360 | |||
U09584.1 | - | 1 | 571 |