Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACFD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135775.2 | NP_001129247.1 | 3 | 381 | ||
NM_001242369.1 | NP_001229298.1 | 4 | 507 | |||
NM_001242370.1 | NP_001229299.1 | 3 | 381 | |||
NM_017586.3 | NP_060056.1 | 4 | 507 | |||
XM_011518181.2 | XP_011516483.1 | 4 | 2064 | |||
XM_011518182.1 | XP_011516484.1 | 4 | 560 | |||
XM_011518184.1 | XP_011516486.1 | 3 | 1123 | |||
XM_011518185.2 | XP_011516487.1 | 4 | 2064 | |||
XM_011518186.1 | XP_011516488.1 | 4 | 560 | |||
GenBank mRNA | AJ011373.1 | - | 4 | 463 | ||
AK074852.1 | - | 4 | 485 | |||
AK075416.1 | - | 3 | 379 | |||
AK075548.1 | - | 4 | 475 | |||
AK296343.1 | - | 4 | 482 | |||
AK298820.1 | - | 3 | 313 | |||
AK304524.1 | - | 5 | 465 | |||
AK311325.1 | - | 4 | 563 | |||
AY358452.1 | - | 4 | 455 | |||
BC030558.1 | - | 4 | 473 | 5223979 | ||
DA699497.1 | - | 3 | 318 | |||
DB165724.1 | - | 4 | 507 |