Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM107A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076778.2 | NP_001070246.1 | 4 | 835 | ||
NM_001282713.1 | NP_001269642.1 | 4 | 635 | |||
NM_001282714.1 | NP_001269643.1 | 5 | 1083 | |||
NM_007177.3 | NP_009108.1 | 5 | 933 | |||
GenBank mRNA | AB023811.1 | - | 4 | 415 | ||
AF035283.1 | - | 4 | 835 | |||
AF089853.1 | - | 4 | 926 | |||
AF089854.1 | - | 5 | 1024 | |||
AK054720.1 | - | 5 | 1083 | |||
AK055443.1 | - | 3 | 971 | |||
AK128791.1 | - | 4 | 462 | |||
AK298001.1 | - | 4 | 590 | |||
AK316450.1 | - | 4 | 490 | |||
AL050264.1 | - | 4 | 448 | |||
BC010561.1 | - | 5 | 534 | 4152118 | ||
BX645008.1 | - | 4 | 646 | |||
CR457133.1 | - | 3 | 374 | |||
EU569833.1 | - | 4 | 490 |