Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LZTS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021020.3 | NP_066300.1 | 3 | 1467 | ||
XM_005273394.3 | XP_005273451.1 | 4 | 1732 | |||
XM_011544383.2 | XP_011542685.1 | 4 | 3670 | |||
XM_011544384.2 | XP_011542686.1 | 4 | 1620 | |||
XM_011544385.2 | XP_011542687.1 | 4 | 1594 | |||
XM_011544386.2 | XP_011542688.1 | 4 | 1819 | |||
GenBank mRNA | AF123655.1 | - | 3 | 1356 | ||
AF123657.1 | - | 3 | 1356 | |||
AF123658.1 | - | 3 | 1356 | |||
AF123659.1 | - | 3 | 1467 | |||
BC075006.2 | - | 3 | 1405 | 30915401 | ||
BC075007.2 | - | 3 | 1405 | 30915614 |