Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161521.1 | NP_001154993.1 | 8 | 938 | ||
NM_001161522.1 | NP_001154994.1 | 7 | 831 | |||
NM_007246.3 | NP_009177.3 | 8 | 1061 | |||
XM_011531572.2 | XP_011529874.1 | 9 | 1051 | |||
XM_011531575.2 | XP_011529877.1 | 7 | 883 | |||
XM_011531576.2 | XP_011529878.1 | 6 | 551 | |||
XM_017007674.1 | XP_016863163.1 | 9 | 1011 | |||
XM_017007675.1 | XP_016863164.1 | 7 | 776 | |||
XM_017007676.1 | XP_016863165.1 | 7 | 884 | |||
XM_017007677.1 | XP_016863166.1 | 6 | 758 | |||
GenBank mRNA | AB209727.1 | - | 4 | 405 | ||
AF059569.1 | - | 8 | 910 | |||
AK025289.1 | - | 5 | 527 | |||
AK294103.1 | - | 8 | 932 | |||
AK294336.1 | - | 4 | 360 | |||
AK308940.1 | - | 4 | 361 | |||
AK315372.1 | - | 8 | 906 | |||
BC022503.1 | - | 8 | 1043 | 4791972 | ||
BC036468.2 | - | 8 | 1002 | 4799622 | ||
BX647776.1 | - | 7 | 815 |