Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SYNRG SIRNA Assays ›
Gene Symbol:
SYNRG
Gene Name:
synergin, gamma
Gene Aliases:
AP1GBP1, SYNG
Chromosome Location:
Chr.17: 37514797 - 37613637 on Build GRCh38
UniGene ID:
Hs.594647
Species:
Human
siRNA ID
HSS117533
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SYNRG
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001163544.2
NP_001157016.1
16, 17
3266
NM_001163545.2
NP_001157017.1
16, 17
3263
NM_001163546.2
NP_001157018.1
16, 17
3131
NM_001163547.2
NP_001157019.1
15, 16
2882
NM_007247.5
NP_009178.3
17, 18
3500
NM_080550.4
NP_542117.3
16, 17
3266
NM_198882.2
NP_942583.1
16, 17
3266
XM_005256980.4
XP_005257037.1
18, 19
3861
XM_017024076.1
XP_016879565.1
18, 19
3861
XM_017024077.1
XP_016879566.1
18, 19
3780
XM_017024078.1
XP_016879567.1
18, 19
3861
XM_017024079.1
XP_016879568.1
18, 19
3798
XM_017024080.1
XP_016879569.1
18, 19
7913
XM_017024081.1
XP_016879570.1
18, 19
7910
XM_017024082.1
XP_016879571.1
18, 19
3861
XM_017024083.1
XP_016879572.1
18, 19
3726
XM_017024084.1
XP_016879573.1
18, 19
3723
XM_017024085.1
XP_016879574.1
18, 19
3798
XM_017024086.1
XP_016879575.1
18, 19
3726
XM_017024087.1
XP_016879576.1
18, 19
3723
XM_017024088.1
XP_016879577.1
17, 18
3627
XM_017024089.1
XP_016879578.1
18, 19
3726
XM_017024090.1
XP_016879579.1
17, 18
3558
XM_017024091.1
XP_016879580.1
17, 18
3477
XM_017024092.1
XP_016879581.1
18, 19
3660
XM_017024093.1
XP_016879582.1
17, 18
3558
XM_017024094.1
XP_016879583.1
17, 18
3477
XM_017024095.1
XP_016879584.1
17, 18
3495
XM_017024096.1
XP_016879585.1
18, 19
3861
XM_017024097.1
XP_016879586.1
17, 18
3423
XM_017024098.1
XP_016879587.1
17, 18
3420
XM_017024099.1
XP_016879588.1
16, 17
3243
XM_017024100.1
XP_016879589.1
17, 18
3420
XM_017024101.1
XP_016879590.1
17, 18
3339
XM_017024102.1
XP_016879591.1
16, 17
3243
XM_017024103.1
XP_016879592.1
16, 17
3186
XM_017024104.1
XP_016879593.1
16, 17
3186
XM_017024106.1
XP_016879595.1
16, 17
3189
GenBank mRNA
AF169548.1
-
17, 18
3460
AK025700.1
-
4, 5
1513
BC090930.1
-
16, 17
3217
3077104
BC117313.1
-
16, 17
3206
40125864
BC143476.1
-
15, 16
2825
9051984
BC143478.1
-
16, 17
3209
9051986
BG120023.1
-
3, 4
317
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
intracellular protein transport
endocytosis
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-r4vqj:80/10.239.163.135:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0