Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001279353.1 | NP_001266282.1 | 2 | 342 | ||
NM_001279354.1 | NP_001266283.1 | 2 | 331 | |||
NM_007259.4 | NP_009190.2 | 2 | 686 | |||
NR_103998.1 | 2 | 311 | ||||
XM_017000164.1 | XP_016855653.1 | 2 | 236 | |||
GenBank mRNA | AF165513.1 | - | 2 | 166 | ||
AJ133421.1 | - | 2 | 154 | |||
AK023170.1 | - | 2 | 173 | |||
AK223214.1 | - | 2 | 139 | |||
AK293555.1 | - | 2 | 172 | |||
AK295529.1 | - | 2 | 186 | |||
AK298786.1 | - | 2 | 253 | |||
AK301994.1 | - | 2 | 293 | |||
AK307949.1 | - | 2 | 132 | |||
BC012932.1 | - | 2 | 172 | 3853152 | ||
BC028382.2 | - | 2 | 667 | 4818323 | ||
BG176899.1 | - | 2 | 342 | |||
DA374440.1 | - | 2 | 311 | |||
U35246.1 | - | 2 | 164 |