Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FKBP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284341.1 | NP_001271270.1 | 5, 6 | 1027 | ||
NM_007270.4 | NP_009201.2 | 4, 5 | 868 | |||
XM_011515115.2 | XP_011513417.1 | 4, 5 | 868 | |||
GenBank mRNA | AF089745.1 | - | 4, 5 | 539 | ||
AK075331.1 | - | 4, 5 | 832 | |||
AK128597.1 | - | 5, 6 | 809 | |||
AK293954.1 | - | 3, 4 | 441 | |||
AK294266.1 | - | 3, 4 | 652 | |||
AK300328.1 | - | 5, 6 | 990 | |||
BC101723.1 | - | 4, 5 | 796 | 8069229 | ||
BC112053.1 | - | 4, 5 | 796 | 8327521 |