Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014891.6 | NP_055706.1 | 2 | 188 | ||
GenBank mRNA | BC000684.2 | - | 2 | 100 | 3347335 | |
BC007873.2 | - | 2 | 64 | 4303959 | ||
BC071993.1 | - | 2 | 98 | |||
BF037671.1 | - | 2 | 133 | |||
BG281467.1 | - | 2 | 188 | |||
BI460344.1 | - | 2 | 179 | |||
U41745.1 | - | 2 | 75 |