Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXOSC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181503.2 | NP_852480.1 | 7 | 635 | ||
XM_006719763.1 | XP_006719826.1 | 6 | 766 | |||
GenBank mRNA | AF025438.1 | - | 6 | 364 | ||
AF279901.1 | - | 8 | 426 | |||
AK096810.1 | - | 5 | 2275 | |||
AK126602.1 | - | 1 | 820 | |||
AL050353.1 | - | 6 | 1858 | |||
BC020773.1 | - | 7 | 396 | 4808747 |