Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014280.2 | NP_055095.2 | 7 | 547 | ||
GenBank mRNA | AF083190.1 | - | 7 | 554 | ||
AF161433.1 | - | 4 | 231 | |||
AF161449.1 | - | 3 | 131 | |||
AK300797.1 | - | 7 | 536 | |||
BC033159.1 | - | 7 | 532 | 4330676 | ||
BX538122.1 | - | 8 | 664 | |||
CR456900.1 | - | 7 | 547 | |||
DB294321.1 | - | 7 | 547 |