Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other R3HDM2 SIRNA Assays ›
Gene Symbol:
R3HDM2
Gene Name:
R3H domain containing 2
Gene Aliases:
CAG6, PR01365
Chromosome Location:
Chr.12: 57253764 - 57430999 on Build GRCh38
UniGene ID:
Hs.443673
Species:
Human
siRNA ID
HSS117766
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: R3HDM2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_014925.3
NP_055740.3
13, 14
1584
XM_005268725.3
XP_005268782.1
15, 16
1934
XM_011538031.1
XP_011536333.1
16, 17
1822
XM_011538033.1
XP_011536335.1
17, 18
1876
XM_011538035.1
XP_011536337.1
16, 17
1780
XM_011538036.1
XP_011536338.1
16, 17
1774
XM_011538037.1
XP_011536339.1
15, 16
1756
XM_011538038.1
XP_011536340.1
15, 16
1726
XM_011538039.1
XP_011536341.1
15, 16
1720
XM_011538040.1
XP_011536342.1
16, 17
1774
XM_011538042.2
XP_011536344.1
18, 19
1972
XM_017018998.1
XP_016874487.1
15, 16
1798
XM_017018999.1
XP_016874488.1
18, 19
2013
XM_017019000.1
XP_016874489.1
16, 17
1780
XM_017019001.1
XP_016874490.1
17, 18
2086
XM_017019002.1
XP_016874491.1
15, 16
1750
XM_017019003.1
XP_016874492.1
15, 16
1726
XM_017019004.1
XP_016874493.1
15, 16
1720
XM_017019005.1
XP_016874494.1
17, 18
1931
XM_017019006.1
XP_016874495.1
16, 17
1824
XM_017019007.1
XP_016874496.1
16, 17
1859
XM_017019008.1
XP_016874497.1
14, 15
1624
XM_017019009.1
XP_016874498.1
15, 16
1678
XM_017019010.1
XP_016874499.1
15, 16
1752
XM_017019011.1
XP_016874500.1
18, 19
1975
XM_017019012.1
XP_016874501.1
14, 15
1660
XM_017019013.1
XP_016874502.1
15, 16
1794
XM_017019014.1
XP_016874503.1
15, 16
1829
XM_017019015.1
XP_016874504.1
15, 16
1971
XM_017019016.1
XP_016874505.1
16, 17
1990
XM_017019017.1
XP_016874506.1
15, 16
1752
XM_017019018.1
XP_016874507.1
15, 16
1718
XM_017019019.1
XP_016874508.1
16, 17
1814
XM_017019020.1
XP_016874509.1
13, 14
1564
XM_017019021.1
XP_016874510.1
15, 16
1788
XM_017019022.1
XP_016874511.1
14, 15
1698
XM_017019023.1
XP_016874512.1
14, 15
1709
XM_017019024.1
XP_016874513.1
14, 15
1698
XM_017019025.1
XP_016874514.1
14, 15
1663
XM_017019026.1
XP_016874515.1
15, 16
1703
XM_017019027.1
XP_016874516.1
14, 15
1834
XM_017019028.1
XP_016874517.1
13, 14
1596
XM_017019029.1
XP_016874518.1
13, 14
1682
XM_017019030.1
XP_016874519.1
6, 7
963
XM_017019031.1
XP_016874520.1
9, 10
1247
XM_017019032.1
XP_016874521.1
5, 6
815
XM_017019033.1
XP_016874522.1
7, 8
966
GenBank mRNA
AB023219.1
-
15, 16
1941
AK293394.1
-
5, 6
836
AX775761.1
-
15, 16
1941
AX775763.1
-
15, 16
1983
BC041857.1
-
5, 6
1396
5270511
BC104995.1
-
3, 4
591
8143998
BC112226.1
-
3, 4
591
8327694
BC143605.1
-
4, 5
693
9052118
Back To Top
More Information
Gene Ontology Categories:
Process(es)
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-d4wl8:80/10.239.162.132:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0