Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other R3HDM2 SIRNA Assays ›
Gene Symbol:
R3HDM2
Gene Name:
R3H domain containing 2
Gene Aliases:
CAG6, PR01365
Chromosome Location:
Chr.12: 57253764 - 57430999 on Build GRCh38
UniGene ID:
Hs.443673
Species:
Human
siRNA ID
HSS117767
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: R3HDM2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_014925.3
NP_055740.3
14, 15
1709
XM_005268725.3
XP_005268782.1
16, 17
2059
XM_011538031.1
XP_011536333.1
17, 18
1947
XM_011538033.1
XP_011536335.1
18, 19
2001
XM_011538035.1
XP_011536337.1
17, 18
1905
XM_011538036.1
XP_011536338.1
17, 18
1899
XM_011538037.1
XP_011536339.1
16, 17
1881
XM_011538038.1
XP_011536340.1
16, 17
1851
XM_011538039.1
XP_011536341.1
16, 17
1845
XM_011538040.1
XP_011536342.1
17, 18
1899
XM_011538042.2
XP_011536344.1
19, 20
2097
XM_017018998.1
XP_016874487.1
16, 17
1923
XM_017018999.1
XP_016874488.1
19, 20
2138
XM_017019000.1
XP_016874489.1
17, 18
1905
XM_017019001.1
XP_016874490.1
18, 19
2211
XM_017019002.1
XP_016874491.1
16, 17
1875
XM_017019003.1
XP_016874492.1
16, 17
1851
XM_017019004.1
XP_016874493.1
16, 17
1845
XM_017019005.1
XP_016874494.1
18, 19
2056
XM_017019006.1
XP_016874495.1
17, 18
1949
XM_017019007.1
XP_016874496.1
17, 18
1984
XM_017019008.1
XP_016874497.1
15, 16
1749
XM_017019009.1
XP_016874498.1
16, 17
1803
XM_017019010.1
XP_016874499.1
16, 17
1877
XM_017019011.1
XP_016874500.1
19, 20
2100
XM_017019012.1
XP_016874501.1
15, 16
1785
XM_017019013.1
XP_016874502.1
16, 17
1919
XM_017019014.1
XP_016874503.1
16, 17
1954
XM_017019015.1
XP_016874504.1
16, 17
2096
XM_017019016.1
XP_016874505.1
17, 18
2115
XM_017019017.1
XP_016874506.1
16, 17
1877
XM_017019018.1
XP_016874507.1
16, 17
1843
XM_017019019.1
XP_016874508.1
17, 18
1939
XM_017019020.1
XP_016874509.1
14, 15
1689
XM_017019021.1
XP_016874510.1
16, 17
1913
XM_017019022.1
XP_016874511.1
15, 16
1823
XM_017019023.1
XP_016874512.1
15, 16
1834
XM_017019024.1
XP_016874513.1
15, 16
1823
XM_017019025.1
XP_016874514.1
15, 16
1788
XM_017019026.1
XP_016874515.1
16, 17
1828
XM_017019027.1
XP_016874516.1
15, 16
1959
XM_017019028.1
XP_016874517.1
14, 15
1721
XM_017019029.1
XP_016874518.1
14, 15
1807
XM_017019030.1
XP_016874519.1
7, 8
1088
XM_017019031.1
XP_016874520.1
10, 11
1372
XM_017019032.1
XP_016874521.1
6, 7
940
XM_017019033.1
XP_016874522.1
8, 9
1091
GenBank mRNA
AB023219.1
-
16, 17
2066
AK293394.1
-
6, 7
961
AX775761.1
-
16, 17
2066
AX775763.1
-
16, 17
2108
BC041857.1
-
6, 7
1521
5270511
BC104995.1
-
4, 5
716
8143998
BC112226.1
-
4, 5
716
8327694
BC143605.1
-
5, 6
818
9052118
Back To Top
More Information
Gene Ontology Categories:
Process(es)
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6d74b6fc5b-xzndl:80/10.239.164.225:80.
git-commit: 0683b6a9b3dadc77a268b704485bdfd9f1fd09b0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.0-2024.10.41-1.0