Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other R3HDM2 SIRNA Assays ›
Gene Symbol:
R3HDM2
Gene Name:
R3H domain containing 2
Gene Aliases:
CAG6, PR01365
Chromosome Location:
Chr.12: 57253764 - 57430999 on Build GRCh38
UniGene ID:
Hs.443673
Species:
Human
siRNA ID
HSS117768
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: R3HDM2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_014925.3
NP_055740.3
18
2163
XM_005268725.3
XP_005268782.1
20
2513
XM_011538031.1
XP_011536333.1
21
2401
XM_011538033.1
XP_011536335.1
22
2455
XM_011538035.1
XP_011536337.1
21
2359
XM_011538036.1
XP_011536338.1
21
2353
XM_011538037.1
XP_011536339.1
20
2335
XM_011538038.1
XP_011536340.1
20
2305
XM_011538039.1
XP_011536341.1
20
2299
XM_011538040.1
XP_011536342.1
21
2353
XM_011538042.2
XP_011536344.1
23
2551
XM_017018998.1
XP_016874487.1
20
2377
XM_017018999.1
XP_016874488.1
23
2592
XM_017019000.1
XP_016874489.1
21
2359
XM_017019001.1
XP_016874490.1
22
2665
XM_017019002.1
XP_016874491.1
20
2329
XM_017019003.1
XP_016874492.1
20
2305
XM_017019004.1
XP_016874493.1
20
2299
XM_017019005.1
XP_016874494.1
22
2510
XM_017019006.1
XP_016874495.1
21
2403
XM_017019007.1
XP_016874496.1
21
2438
XM_017019008.1
XP_016874497.1
19
2203
XM_017019009.1
XP_016874498.1
20
2257
XM_017019010.1
XP_016874499.1
20
2331
XM_017019011.1
XP_016874500.1
23
2554
XM_017019012.1
XP_016874501.1
19
2239
XM_017019013.1
XP_016874502.1
20
2373
XM_017019014.1
XP_016874503.1
20
2408
XM_017019015.1
XP_016874504.1
20
2550
XM_017019016.1
XP_016874505.1
21
2569
XM_017019017.1
XP_016874506.1
20
2331
XM_017019018.1
XP_016874507.1
20
2297
XM_017019019.1
XP_016874508.1
21
2393
XM_017019020.1
XP_016874509.1
18
2143
XM_017019021.1
XP_016874510.1
20
2367
XM_017019022.1
XP_016874511.1
19
2277
XM_017019023.1
XP_016874512.1
19
2288
XM_017019024.1
XP_016874513.1
19
2277
XM_017019025.1
XP_016874514.1
19
2242
XM_017019026.1
XP_016874515.1
20
2282
XM_017019027.1
XP_016874516.1
19
2413
XM_017019028.1
XP_016874517.1
18
2175
XM_017019029.1
XP_016874518.1
18
2261
XM_017019030.1
XP_016874519.1
11
1542
XM_017019031.1
XP_016874520.1
14
1826
XM_017019032.1
XP_016874521.1
10
1394
XM_017019033.1
XP_016874522.1
12
1545
GenBank mRNA
AB023219.1
-
20
2520
AK293394.1
-
10
1415
AX775761.1
-
20
2520
AX775763.1
-
20
2562
BC041857.1
-
10
1975
5270511
BC104995.1
-
8
1170
8143998
BC112226.1
-
8
1170
8327694
BC143605.1
-
9
1272
9052118
Back To Top
More Information
Gene Ontology Categories:
Process(es)
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-6lbnp:80/10.239.161.162:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline