Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNKSR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168647.1 | NP_001162118.1 | 19 | 2553 | ||
NM_001168648.1 | NP_001162119.1 | 20 | 2643 | |||
NM_001168649.1 | NP_001162120.1 | 19 | 2496 | |||
NM_014927.3 | NP_055742.2 | 20 | 2643 | |||
XM_011545471.2 | XP_011543773.1 | 19 | 2636 | |||
XM_011545472.2 | XP_011543774.1 | 19 | 2500 | |||
XM_017029358.1 | XP_016884847.1 | 18 | 2410 | |||
GenBank mRNA | AB020709.1 | - | 20 | 2619 | ||
AF418269.1 | - | 20 | 2199 | |||
AF418270.1 | - | 20 | 2199 | |||
AK056549.1 | - | 9 | 995 | |||
AK294728.1 | - | 19 | 2458 | |||
BC126121.1 | - | 20 | 2198 | 8991837 | ||
BC136289.1 | - | 20 | 2198 | 9020276 | ||
BC143839.1 | - | 19 | 2108 | 9052354 |