Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNKSR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168647.1 | NP_001162118.1 | 9 | 1340 | ||
NM_001168648.1 | NP_001162119.1 | 9 | 1340 | |||
NM_014927.3 | NP_055742.2 | 9 | 1340 | |||
XM_011545471.2 | XP_011543773.1 | 9 | 1423 | |||
GenBank mRNA | AB020709.1 | - | 9 | 1316 | ||
AF418269.1 | - | 9 | 896 | |||
AF418270.1 | - | 9 | 896 | |||
BC126121.1 | - | 9 | 895 | 8991837 | ||
BC136289.1 | - | 9 | 895 | 9020276 | ||
BC143839.1 | - | 9 | 895 | 9052354 |